missing translation for 'onlineSavingsMsg'
Få mere at vide

(R)-(+)-beta-methylphenethylamine, TRC

Artikelnummer. 30343281
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343281 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30343281 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3999381G
 Begrænset vare

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (R)-(+)-beta-methylphenethylamine
CAS 28163-64-6
Molekylær formel C9 H13 N
Mængde 1 g
Synonym (βR)-β-Methylbenzeneethanamine, ((R)-2-Phenylpropan-1-yl)amine, (+)-2-Phenylpropylamine, (+)-β-Methylphenethylamine, (2R)-1-Amino-2-phenylpropane, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropylamine, (R)-(+)--Methylphenethylamine, (R)-(+)-2-Phenylpropylamine, (R)-2-Phenyl-1-propanamine, (R)-2-Phenyl-1-propylamine, (R)-2-Phenylpropylamine, (R)-2-Phenylpropylamine, (R)-β-Methylphenethylamine
InChI formel InChI=1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m0/s1
SMIL C[C@@H](CN)c1ccccc1
IUPAC navn (2R)-2-phenylpropan-1-amin
Molekylvægt (g/mol) 135.21
Formel vægt 135.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.