Få mere at vide
(R)-Baclofen, TRC
Artikelnummer. 30137803
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137803 25 mg
30132873 10 mg
30012303 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137803 Leverandør Toronto Research Chemicals Leverandørnr. TRCB10799025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 69308-37-8
Molekylær formel C10 H12 Cl N O2
Molekylvægt (g/mol) 213.66
Synonym (βR)-β-(Aminomethyl)-4-chlorobenzenepropanoic Acid, (-)-Baclofen, (R)-(-)-Baclofen, (R)-4-Amino-3-(4-chlorophenyl)butanoic Acid, (R)-4-Amino-3-(4-chlorophenyl)butyric Acid, D-Baclofen, R-(-)-Baclofen, l-Baclofen, AGI 006, Arbaclofen
IUPAC navn (3R)-4-amino-3-(4-chlorophenyl)butanoic acid
SMIL NC[C@H](CC(=O)O)c1ccc(Cl)cc1
Tekniske data
Kemisk navn eller materiale (R)-Baclofen
InChI formel InChI=1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)/t8-/m0/s1
Mængde 25 mg
Formel vægt 213.0557