Få mere at vide
(R)-Apomorphine Hydrochloride Hemihydrate, TRC
Artikelnummer. 30138757
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138757 50 mg
30136494 25 mg
30004593 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138757 Leverandør Toronto Research Chemicals Leverandørnr. TRCA72750050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 41372-20-7
Molekylær formel 2 C17 H17 N O2 . 2 Cl H . H2 O
Molekylvægt (g/mol) 625.58
Synonym 4H-Dibenzo[de,g]quinoline-10,11-diol, 5,6,6a,7-tetrahydro-6-methyl-, hydrochloride, hydrate (2:2:1), (6aR)-, 4H-Dibenzo[de,g]quinoline-10,11-diol, 5,6,6a,7-tetrahydro-6-methyl-, hydrochloride, hydrate (2:1), (6aR)- (9CI), 4H-Dibenzo[de,g]quinoline-10,11-diol, 5,6,6a,7-tetrahydro-6-methyl-, hydrochloride, hydrate (2:1), (R)-, (R)-(-)-Apomorphine hydrochloride hemihydrate, Apomorphine hydrochloride hemihydrate, (R)-6-Methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-10,11-diol hydrochloride hemihydrate
IUPAC navn (6aR)-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-10,11-diol;hydrate;dihydrochloride
SMIL O.Cl.Cl.CN1CCc2cccc3c2[C@H]1Cc4ccc(O)c(O)c34.CN5CCc6cccc7c6[C@H]5Cc8ccc(O)c(O)c78
Tekniske data
Kemisk navn eller materiale Apomorphine Hydrochloride Hemihydrate
Analyt- eller komponentnavne Apomorphine Hydrochloride Hemihydrate
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/2C17H17NO2.2ClH.H2O/c2*1-18-8-7-10-3-2-4-12-15(10)13(18)9-11-5-6-14(19)17(20)16(11)12;;;/h2*2-6,13,19-20H,7-9H2,1H3;2*1H;1H2/t2*13-;;;/m11.../s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 50 mg
Formel vægt 624.2158
Procent renhed >95%