Få mere at vide
(R)-Apomorphine-d4 Hydrochloride, TRC
Artikelnummer. 30240689
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240689 1 mg
30240974 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240689 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7275011MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76787-63-8
Molekylær formel C17H13D4NO2 . HCl
Molekylvægt (g/mol) 308.8
Synonym (R)-5,6,6a,7-Tetrahydro-6-methyl-4H-dibenzo[de,g]quinoline-1,2,3,8,9-d5-10,11-diol-d5 Hydrochloride-d4, Apomorphine-1,2,3,8,9-d5 Hydrochloride-d4;
IUPAC navn (6aR)-1,2,3,8,9-pentadeuterio-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-10,11-diol;hydrochloride
SMIL Cl.[2H]c1cc([2H])c2c3[C@@H](Cc4c([2H])c([2H])c(O)c(O)c24)N(C)CCc13
Tekniske data
Kemisk navn eller materiale (R)-Apomorphine-d4 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C17H17NO2.ClH/c1-18-8-7-10-3-2-4-12-15(10)13(18)9-11-5-6-14(19)17(20)16(11)12;/h2-6,13,19-20H,7-9H2,1H3;1H/t13-;/m1./s1/i2D,3D,4D,5D,6D;
Formel vægt 307.1277