Få mere at vide
(R)-Amino Carnitine, TRC
Artikelnummer. 30311282
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311282 10 mg
30315441 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30311282 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60340010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Amino Carnitine
CAS 98063-21-9
Renhedsgrad noter HPLC
Molekylær formel C7 H16 N2 O2
Mængde 10 mg
Synonym 1-Propanaminium, 2-amino-3-carboxy-N,N,N-trimethyl-, inner salt, (2R)-, 1-Propanaminium, 2-amino-3-carboxy-N,N,N-trimethyl-, inner salt, (R)-, (R)-3-Amino-4-(trimethylammonio)butyrate, Emeriamine, (R)-Aminocarnitine
InChI formel InChI=1S/C7H16N2O2/c1-9(2,3)5-6(8)4-7(10)11/h6H,4-5,8H2,1-3H3/t6-/m1/s1
SMIL C[N+](C)(C)C[C@H](N)CC(=O)[O-]
IUPAC navn (3R)-3-amino-4-(trimethylazaniumyl)butanoate
Molekylvægt (g/mol) 160.21
Formel vægt 160.1212
Procent renhed >95