Få mere at vide
(R)-Alpha-allyl-proline hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (R)-alpha-allyl-proline hydrochloride |
| CAS | 177206-69-8 |
| Molekylær formel | C8H13NO2 . HCl |
| Mængde | 10 mg |
| Synonym | L-Proline, 2-(2-propenyl)-, hydrochloride (9CI), (R)-2-Allyl-2-pyrrolidinecarboxylic acid hydrochloride, 2-(2-Propen-1-yl)-L-proline hydrochloride, (2R)-2-(Prop-2-enyl)pyrrolidine-2-carboxylic acid hydrochloride, α-Allyl-L-proline hydrochloride, (R)-α-Allyl-proline hydrochloride |
| InChI formel | InChI=1S/C8H13NO2.ClH/c1-2-4-8(7(10)11)5-3-6-9-8;/h2,9H,1,3-6H2,(H,10,11);1H/t8-;/m0./s1 |
| SMIL | Cl.OC(=O)[C@]1(CC=C)CCCN1 |
| IUPAC navn | (2R)-2-prop-2-enylpyrrolidine-2-carboxylic acid;hydrochloride |
| Molekylvægt (g/mol) | 191.65 |
| Formel vægt | 191.0713 |