Få mere at vide
(R)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid, TRC
Artikelnummer. 30371678
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371678 1 g
30376994 25 g
30371796 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30371678 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9169701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63422-71-9
Molekylær formel C15 H17 N3 O5
Molekylvægt (g/mol) 319.31
Synonym (2R)-2-[[(4-Ethyl-2,3-dioxopiperazin-1-yl)carbonyl]amino]-2-phenylacetic Acid, Piperacillin Sodium Imp. G (EP)
IUPAC navn (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetic acid
SMIL CCN1CCN(C(=O)N[C@@H](C(=O)O)c2ccccc2)C(=O)C1=O
Tekniske data
Kemisk navn eller materiale (2R)-2-[((4-Ethyl-2,3-dioxopiperazin-1-yl)carbonyl)amino]-2-phenylacetic Acid
Mængde 1 g
InChI formel InChI=1S/C15H17N3O5/c1-2-17-8-9-18(13(20)12(17)19)15(23)16-11(14(21)22)10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3,(H,16,23)(H,21,22)/t11-/m1/s1
Formel vægt 319.1168