Få mere at vide
(R)-9-(2-Hydroxypropyl)adenine (Desphosphoryltenofovir), Mikromol™
Artikelnummer. 30051487
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051487 25 mg
30049485 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051487 Leverandør Mikromol Leverandørnr. MM1329.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(R)-9-(2-Hydroxypropyl)adenine (Desphosphoryltenofovir)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-9-(2-Hydroxypropyl)adenine
CAS 14047-28-0
Analyt- eller komponentnavne (R)-9-(2-Hydroxypropyl)adenine
Forsendelsestilstand Room Temperature
Molekylær formel C8 H11 N5 O
Certificeringer/compliance ISO 17025
Synonym (R)-9-(2-Hydroxypropyl)adenine, 9-[(2R)-2-Hydroxypropyl]adenine, 9-[2-(R)-(Hydroxy)propyl]adenine, D-(-)-9-(2-Hydroxypropyl)adenine, (alphaR)-6-Amino-alpha-methyl-9H-purine-9-ethanol, 9H-Purine-9-ethanol, 6-amino-alpha-methyl-, (R)-, 9H-Purine-9-ethanol, 6-amino-alpha-methyl-, D- (8CI)
InChI formel InChI=1S/C8H11N5O/c1-5(14)2-13-4-12-6-7(9)10-3-11-8(6)13/h3-5,14H,2H2,1H3,(H2,9,10,11)/t5-/m1/s1
SMIL C[C@@H](O)Cn1cnc2c(N)ncnc12
IUPAC navn (2R)-1-(6-aminopurin-9-yl)propan-2-ol
Molekylvægt (g/mol) 193.21
Mængde 25 mg
Formel vægt 193.0964 g/mol