missing translation for 'onlineSavingsMsg'
Få mere at vide

R 84852, TRC

Artikelnummer. 30420810
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420810 5 mg
30412282 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30420810 Leverandør Toronto Research Chemicals Leverandørnr. TRCR0701305MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 152542-03-5
Molekylær formel C23 H28 F N3 O4
Molekylvægt (g/mol) 429.48
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(4-fluoro-2-hydroxybenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-, 3-[2-[4-(4-Fluoro-2-hydroxybenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, R 84852, 3-[2-[4-(4-Fluoro-2-hydroxybenzoyl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
IUPAC navn 3-[2-[4-(4-fluoro-2-hydroxybenzoyl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMIL CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3O)C(=O)N4CCCC(O)C4=N1

Tekniske data

Kemisk navn eller materiale R 84852
Mængde 5 mg
InChI formel InChI=1S/C23H28FN3O4/c1-14-17(23(31)27-9-2-3-19(28)22(27)25-14)8-12-26-10-6-15(7-11-26)21(30)18-5-4-16(24)13-20(18)29/h4-5,13,15,19,28-29H,2-3,6-12H2,1H3
Formel vægt 429.2064
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.