missing translation for 'onlineSavingsMsg'
Få mere at vide

(R)-6-Bromo Phenylephrine Hydrochloride, TRC

Artikelnummer. 30333432
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30333432 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30333432 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6868301MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (R)-6-Bromo Phenylephrine Hydrochloride
CAS 1391067-95-0
Molekylær formel C9H13BrClNO2
Mængde 1 mg
Synonym (R)-6-Bromo-3-hydroxy-alpha-[(methylamino)methyl]benzenemethanol Hydrochloride, (R)-6-Bromophenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-9(13)7-4-6(12)2-3-8(7)10;/h2-4,9,11-13H,5H2,1H3;1H/t9-;/m0./s1
SMIL Cl.CNC[C@H](O)c1cc(O)ccc1Br
IUPAC navn 4-bromo-3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvægt (g/mol) 282.56
Formel vægt 280.9818
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.