missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7054605
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
114360-56-4
C24H28N2O6
440.496
MFCD00798647
LIWKOFAHRLBNMG-HXUWFJFHSA-N
fmoc-d-dab boc-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, n-a-fmoc-n-g-boc-d-2,4-diaminobutyric acid, 2r-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-t-butyloxycarbonyl-d-2,4-diaminobutyric acid, ambotzfaa1317, fmoc-4-boc-d-2,4-diaminobutyric acid, n-, a-fmoc-n-, a-boc-d-2,4-diaminobutyricacid, n-a-fmoc-n-gamma-boc-d-2,4-diaminobutyric acid, 2r-4-boc-amino-2-fmoc-amino-butanoic acid
7021129
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
114360-56-4
C24H28N2O6
440.496
MFCD00798647
LIWKOFAHRLBNMG-HXUWFJFHSA-N
fmoc-d-dab boc-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, n-a-fmoc-n-g-boc-d-2,4-diaminobutyric acid, 2r-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-t-butyloxycarbonyl-d-2,4-diaminobutyric acid, ambotzfaa1317, fmoc-4-boc-d-2,4-diaminobutyric acid, n-, a-fmoc-n-, a-boc-d-2,4-diaminobutyricacid, n-a-fmoc-n-gamma-boc-d-2,4-diaminobutyric acid, 2r-4-boc-amino-2-fmoc-amino-butanoic acid
7021129
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 440.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'