Learn More
Alfa Aesar™ (R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%
Brand: Alfa Aesar™ H62817.MD
Additional Details : CAS Number : 101854-42-6 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
101854-42-6 | |
533.71 | |
CBSVEVKFQHTZSP-BTQNPOSSSA-N | |
45357174 | |
CC(C)(C)OC(=O)NC(CCNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
C29H47N3O6 | |
MFCD00798628 | |
boc-d-dab z-oh.dcha, dicyclohexylamine r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoate, boc-d-dab z-oh*dcha, n-boc-n'-cbz-d-2,4-diaminobutyric acid dicyclohexylamine salt, r-2-1,1-dimethylethoxy carbonyl amino-4-phenylmethoxy carbonyl amino butanoic acid, dicyclohexylamine, n-a-boc-n-?-z-d-2,4-diaminobutyric aciddicyclohexylaminesalt, 2r-4-benzyloxycarbonylamino-2-tert-butoxycarbonylamino butanoic acid; n-cyclohexylcyclohexanamine, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid; dicha | |
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
(R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt | |
101854-42-6 | |
MFCD00798628 | |
CBSVEVKFQHTZSP-BTQNPOSSSA-N | |
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-(phenylmethoxycarbonylamino)butanoic acid | |
45357174 | |
Solid |
250mg | |
C29H47N3O6 | |
boc-d-dab z-oh.dcha, dicyclohexylamine r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoate, boc-d-dab z-oh*dcha, n-boc-n'-cbz-d-2,4-diaminobutyric acid dicyclohexylamine salt, r-2-1,1-dimethylethoxy carbonyl amino-4-phenylmethoxy carbonyl amino butanoic acid, dicyclohexylamine, n-a-boc-n-?-z-d-2,4-diaminobutyric aciddicyclohexylaminesalt, 2r-4-benzyloxycarbonylamino-2-tert-butoxycarbonylamino butanoic acid; n-cyclohexylcyclohexanamine, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid; dicha | |
CC(C)(C)OC(=O)NC(CCNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2 | |
533.71 | |
533.7 | |
98% |