Få mere at vide
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone, TRC
Artikelnummer. 30333934
Skift visning
Mængde:
1 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30333934 1 g
30341090 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30333934 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2887051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone
CAS 131685-53-5
Molekylær formel C13H15NO3
Mængde 1 g
Synonym (R)-3-(1-Oxopropyl)-4-(phenylmethyl)-2-oxazolidinone, (-)-4-Benzyl-3-propionyl-2-oxazolidinone, (4R)-4-Benzyl-3-propionyloxazolidin-2-one, (4R)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one, (R)-3-(1-Oxopropyl)-4-benzyl-2-oxazolidinone, (R)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one, (R)-4-Benzyl-3-propionyl-oxazolidin-2-one, (R)-4-Benzyl-3-propionyloxazolidin-2-one
InChI formel InChI=1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m1/s1
SMIL CCC(=O)N1[C@H](Cc2ccccc2)COC1=O
IUPAC navn (4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 233.26
Formel vægt 233.1052