Få mere at vide
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone-D3, TRC
Artikelnummer. 30329370
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30329370 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329370 Leverandør Toronto Research Chemicals Leverandørnr. TRCB288706100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone-D3
CAS 1352449-25-2
Molekylær formel C13H12D3NO3
Mængde 100 mg
Synonym (R)-3-(1-Oxopropyl)-4-(phenylmethyl)-2-oxazolidinone-D3, (-)-4-Benzyl-3-propionyl-2-oxazolidinone-D3, (4R)-4-Benzyl-3-propionyloxazolidin-2-one-D3, (4R)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one-D3, (R)-3-(1-Oxopropyl)-4-benzyl-2-oxazolidinone-D3, (R)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one-D3, (R)-4-Benzyl-3-propionyl-oxazolidin-2-one-D3, (R)-4-Benzyl-3-propionyloxazolidin-2-one-D3
InChI formel InChI=1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m1/s1/i1D3
SMIL [2H]C([2H])([2H])CC(=O)N1[C@H](Cc2ccccc2)COC1=O
IUPAC navn (4R)-4-benzyl-3-(3,3,3-trideuteriopropanoyl)-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 236.28
Formel vægt 236.124