missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Piperidinamine dihydrochloride, 97%, Thermo Scientific™

Catalog Number p-4228874
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
334618-23-4
C5H12N2·2ClH
173.09
GGPNYXIOFZLNKW-ZJIMSODOSA-N
r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl
16211333
(3R)-piperidin-3-amine;dihydrochloride
C1CC(CNC1)N.Cl.Cl
This item is not returnable. View return policy
334618-23-4
C5H12N2·2ClH
173.09
GGPNYXIOFZLNKW-ZJIMSODOSA-N
r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl
16211333
(3R)-piperidin-3-amine;dihydrochloride
C1CC(CNC1)N.Cl.Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 207.5°C to 211.5°C
Color Tan to White
Elemental Analysis 33.5 to 35.9% C, 15.5 to 16.8% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Specific Rotation -1° to -3° (20°C, 589nm) (c=1, water)
Formula Weight 173.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(-)-3-Piperidinamine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-3-Piperidinamine dihydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'