missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Phenylmorpholine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7077638
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1624261-27-3
C10H13NO·ClH
199.68
BZPHPXWWVHPSOP-PPHPATTJSA-N
r-3-phenylmorpholine hydrochloride, 3r-3-phenyl-morpholine hcl, 3r-3-phenylmorpholine hydrochloride
71650782
(3R)-3-phenylmorpholine;hydrochloride
C1COCC(N1)C2=CC=CC=C2.Cl
This item is not returnable. View return policy
1624261-27-3
C10H13NO·ClH
199.68
BZPHPXWWVHPSOP-PPHPATTJSA-N
r-3-phenylmorpholine hydrochloride, 3r-3-phenyl-morpholine hcl, 3r-3-phenylmorpholine hydrochloride
71650782
(3R)-3-phenylmorpholine;hydrochloride
C1COCC(N1)C2=CC=CC=C2.Cl
Melting Point 192.0°C to 193.5°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Quantity 1 g
Formula Weight 199.68
Percent Purity 97%
Chemical Name or Material (R)-3-Phenylmorpholine hydrochloride
compliance-icons
Product Identifier
  • (R)-3-Phenylmorpholine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'