missing translation for 'onlineSavingsMsg'
Få mere at vide

(R)-3-Hydroxy Myristic Acid Dicyclohexylammonium Salt, TRC

Artikelnummer. 30399809
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30399809 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399809 Supplier Toronto Research Chemicals Supplier No. TRCH948515500MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (R)-3-Hydroxy Myristic Acid Dicyclohexylammonium Salt
CAS 76062-98-1
Molekylær formel C26H51NO3
Mængde 500 mg
Synonym (R)-3-Hydroxytetradecanoic Acid N-cyclohexylcyclohexanamine Salt, D-3-Hydroxytetradecanoic Acid N-cyclohexylcyclohexanamine Salt
InChI formel InChI=1S/C14H28O3.C12H23N/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h13,15H,2-12H2,1H3,(H,16,17);11-13H,1-10H2/t13-;/m1./s1
SMIL CCCCCCCCCCC[C@H](CC(=O)O)O.C1CCC(CC1)NC2CCCCC2
IUPAC navn N-cyclohexylcyclohexanamine;(3R)-3-hydroxytetradecanoic acid
Molekylvægt (g/mol) 425.69
Formel vægt 425.3869
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.