missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-3-Hydroxy-3-phenylpropionic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 118°C to 121°C |
Quantity | 1 g |
Beilstein | 3197772 |
Optical Rotation | +22° (c=4 in Methanol) |
Formula Weight | 166.18 |
Percent Purity | ≥98% |
Chemical Name or Material | (R)-(+)-3-Hydroxy-3-phenylpropionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'