missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(R)-(-)-2-Phenylglycine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride |
| CAS | 19883-41-1 |
| Molekylær formel | C9H11NO2.HCl |
| Mængde | 5 g |
| Synonym | Benzeneacetic acid','α-amino-','methyl ester','hydrochloride (1:1)','(αR)-, Benzeneacetic acid','α-amino-','methyl ester','hydrochloride','(R)-, Benzeneacetic acid','α-amino-','methyl ester','hydrochloride','(αR)- (9CI), Glycine','2-phenyl-','methyl ester','hydrochloride','D- (8CI), ((R)-2-Methoxy-2-oxo-1-phenylethyl)ammonium chloride, (R)-(-)-2-Phenylglycine methyl ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Phenylglycine methyl ester hydrochloride, D-Phenylglycine methyl ester hydrochloride, D-α-Aminophenylacetic acid methyl ester hydrochloride, Methyl (R)-amino(phenyl)acetate hydrochloride, Methyl (R)-phenylglycinate hydrochloride, Methyl D-phenylglycinate hydrochloride, Methyl D-α-phenylglycinate hydrochloride, Methyl D-α-phenylglycine hydrochloride |
| InChI formel | InChI=1S/C9H11NO2.ClH/c1-12-9(11)8(10)7-5-3-2-4-6-7;/h2-6,8H,10H2,1H3;1H/t8-;/m1./s1 |
| SMIL | Cl.COC(=O)[C@H](N)c1ccccc1 |
| IUPAC navn | methyl (2R)-2-amino-2-phenylacetate;hydrochloride |
| Molekylvægt (g/mol) | 201.65 |
| Formel vægt | 201.0557 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?