missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(R)-(-)-2-Phenylglycine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride |
| CAS | 19883-41-1 |
| Molekylær formel | C9H11NO2.HCl |
| Mængde | 1 g |
| Synonym | Benzeneacetic acid','α-amino-','methyl ester','hydrochloride (1:1)','(αR)-, Benzeneacetic acid','α-amino-','methyl ester','hydrochloride','(R)-, Benzeneacetic acid','α-amino-','methyl ester','hydrochloride','(αR)- (9CI), Glycine','2-phenyl-','methyl ester','hydrochloride','D- (8CI), ((R)-2-Methoxy-2-oxo-1-phenylethyl)ammonium chloride, (R)-(-)-2-Phenylglycine methyl ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Phenylglycine methyl ester hydrochloride, D-Phenylglycine methyl ester hydrochloride, D-α-Aminophenylacetic acid methyl ester hydrochloride, Methyl (R)-amino(phenyl)acetate hydrochloride, Methyl (R)-phenylglycinate hydrochloride, Methyl D-phenylglycinate hydrochloride, Methyl D-α-phenylglycinate hydrochloride, Methyl D-α-phenylglycine hydrochloride |
| InChI formel | InChI=1S/C9H11NO2.ClH/c1-12-9(11)8(10)7-5-3-2-4-6-7;/h2-6,8H,10H2,1H3;1H/t8-;/m1./s1 |
| SMIL | Cl.COC(=O)[C@H](N)c1ccccc1 |
| IUPAC navn | methyl (2R)-2-amino-2-phenylacetate;hydrochloride |
| Molekylvægt (g/mol) | 201.65 |
| Formel vægt | 201.0557 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?