Få mere at vide
(R)-2-Methylbutyric Acid Chloride-d3, TRC
Artikelnummer. 30469930
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30469930 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469930 Leverandør Toronto Research Chemicals Leverandørnr. TRCM294547500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-2-Methylbutyric Acid Chloride-d3
CAS 1795785-64-6
Molekylær formel C17 H18 N4 O . H Cl
Mængde 500 mg
InChI formel InChI=1S/C17H18N4O.ClH/c1-11-13(19-10-18-11)9-21-8-7-15-16(17(21)22)12-5-3-4-6-14(12)20(15)2;/h3-6,10H,7-9H2,1-2H3,(H,18,19);1H
SMIL Cl.Cc1nc[nH]c1CN2CCc3c(C2=O)c4ccccc4n3C
IUPAC navn 5-methyl-2-[(4-methyl-1H-imidazol-5-yl)methyl]-3,4-dihydropyrido[4,3-b]indol-1-one;hydrochloride
Molekylvægt (g/mol) 330.812
Formel vægt 330.125