Få mere at vide
(R)-(-)-2-Chlorophenylglycine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (R)-(-)-2-Chlorophenylglycine Methyl Ester Hydrochloride |
| CAS | 212838-70-5 |
| Molekylær formel | C9H11Cl2NO2 |
| Mængde | 500 mg |
| Synonym | Benzeneacetic acid, α-amino-2-chloro-, methyl ester, hydrochloride (1:1), (αR)- (ACI), Benzeneacetic acid, α-amino-2-chloro-, methyl ester, hydrochloride, (αR)- (9CI), (R)-Methyl 2-amino-2-(2-chlorophenyl)acetate hydrochloride, Methyl (2R)-2-amino-2-(2-chlorophenyl)acetate hydrochloride |
| InChI formel | InChI=1S/C9H10ClNO2.ClH/c1-13-9(12)8(11)6-4-2-3-5-7(6)10;/h2-5,8H,11H2,1H3;1H/t8-;/m1./s1 |
| SMIL | Cl.COC(=O)[C@H](N)c1ccccc1Cl |
| IUPAC navn | methyl (2R)-2-amino-2-(2-chlorophenyl)acetate;hydrochloride |
| Molekylvægt (g/mol) | 236.1 |
| Formel vægt | 235.0167 |