Få mere at vide
(R)-2-Bromo Phenylephrine Hydrochloride, TRC
Artikelnummer. 30343825
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343825 10 mg
30341648 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30343825 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68680510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-2-Bromo Phenylephrine Hydrochloride
CAS 1391054-09-3
Molekylær formel C9H13BrClNO2
Mængde 10 mg
Synonym Rac 2-Bromo Phenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-8(13)6-3-2-4-7(12)9(6)10;/h2-4,8,11-13H,5H2,1H3;1H/t8-;/m0./s1
SMIL Cl.CNC[C@H](O)c1cccc(O)c1Br
IUPAC navn 2-bromo-3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvægt (g/mol) 282.56
Formel vægt 280.9818