missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Benzylamino-1-butanol, 99%, Thermo Scientific Chemicals

Catalog Number 11499490
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
6257-49-4
C11H17NO
179.263
MFCD00085354
PGFBTQBTIYCCFJ-LLVKDONJSA-N
r-2-benzylamino butan-1-ol, 2r-2-benzylamino butan-1-ol, r---2-benzylamino-1-butanol, r-2-benzylamino-1-butanol, 2-benzylamino-1-butanol #, r---n-benzyl-2-amino-1-butanol, 2r-2-phenylmethyl amino-1-butanol, pound r pound c---2-benzylamino butan-1-ol
927306
(2R)-2-(benzylamino)butan-1-ol
CCC(CO)NCC1=CC=CC=C1
This item is not returnable. View return policy
6257-49-4
C11H17NO
179.263
MFCD00085354
PGFBTQBTIYCCFJ-LLVKDONJSA-N
r-2-benzylamino butan-1-ol, 2r-2-benzylamino butan-1-ol, r---2-benzylamino-1-butanol, r-2-benzylamino-1-butanol, 2-benzylamino-1-butanol #, r---n-benzyl-2-amino-1-butanol, 2r-2-phenylmethyl amino-1-butanol, pound r pound c---2-benzylamino butan-1-ol
927306
(2R)-2-(benzylamino)butan-1-ol
CCC(CO)NCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 179.26
Percent Purity 99%
Chemical Name or Material (R)-(-)-2-Benzylamino-1-butanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'