Få mere at vide
(R)-1-Methyl-3-aminomethyl-piperidine Dihydrochloride, TRC
Artikelnummer. 30466391
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466391 100 mg
30477755 250 mg
30475188 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30466391 Leverandør Toronto Research Chemicals Leverandørnr. TRCM221783100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1400744-18-4
Molekylær formel C7H16N2 . 2 (HCl)
Molekylvægt (g/mol) 201.13
Synonym 3-Piperidinemethanamine, 1-methyl-, hydrochloride (1:2), (3R)- (ACI), (3R)-1-Methyl-3-piperidinemethanamine dihydrochloride, (R)-(1-Methylpiperidin-3-yl)methanamine dihydrochloride, [(3R)-1-Methylpiperidin-3-yl]methanamine dihydrochloride, (3R)-3-(Aminomethyl)-1-methylpiperidine dihydrochloride, (3R)-3-(Aminomethyl)-N-methylpiperidine dihydrochloride, (R)-N-Methyl-3-(aminomethyl)piperidine dihydrochloride
IUPAC navn [(3R)-1-methylpiperidin-3-yl]methanamine;dihydrochloride
SMIL Cl.Cl.CN1CCC[C@H](CN)C1
Tekniske data
Kemisk navn eller materiale (R)-1-Methyl-3-aminomethyl-piperidine Dihydrochloride
Mængde 100 mg
InChI formel InChI=1S/C7H16N2.2ClH/c1-9-4-2-3-7(5-8)6-9;;/h7H,2-6,8H2,1H3;2*1H/t7-;;/m1../s1
Formel vægt 200.0847