missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Benzyl-3-hydroxypiperidine, 97%, Thermo Scientific™

Catalog Number 15484017
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
91599-81-4
C12H17NO
191.274
MFCD00274307
UTTCOAGPVHRUFO-GFCCVEGCSA-N
r-1-benzylpiperidin-3-ol, r-1-benzyl-3-hydroxypiperidine, 3r-1-benzylpiperidin-3-ol, r-1-n-benzyl-3-hydroxy-piperidine, r-1-benzyl-3-piperidinol, r---1-benzyl-3-hydroxypiperidine, r-n-benzyl-3-hydroxypiperidine, 3-piperidinol, 1-phenylmethyl-, 3r, r-pound inverted question mark-1-benzyl-3-hydroxypiperidinol, pubchem13101
693761
(3R)-1-benzylpiperidin-3-ol
C1CC(CN(C1)CC2=CC=CC=C2)O
This item is not returnable. View return policy
91599-81-4
C12H17NO
191.274
MFCD00274307
UTTCOAGPVHRUFO-GFCCVEGCSA-N
r-1-benzylpiperidin-3-ol, r-1-benzyl-3-hydroxypiperidine, 3r-1-benzylpiperidin-3-ol, r-1-n-benzyl-3-hydroxy-piperidine, r-1-benzyl-3-piperidinol, r---1-benzyl-3-hydroxypiperidine, r-n-benzyl-3-hydroxypiperidine, 3-piperidinol, 1-phenylmethyl-, 3r, r-pound inverted question mark-1-benzyl-3-hydroxypiperidinol, pubchem13101
693761
(3R)-1-benzylpiperidin-3-ol
C1CC(CN(C1)CC2=CC=CC=C2)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-Benzyl-3-hydroxypiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'