Få mere at vide
Quizalofop-p-tefuryl, TRC
Artikelnummer. 30008106
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008106 50 mg
30009080 100 mg
30011728 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008106 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ82611850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 119738-06-6
Molekylær formel C22 H21 Cl N2 O5
Molekylvægt (g/mol) 428.8655
Synonym Propanoic acid, 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-, (tetrahydro-2-furanyl)methyl ester, Quizalofop-tefuryl, UBI 1956, (±) tetrahydrofurfuryl (R)-2-[4-(6-chloroquinoxalin-2-yloxy)phenyloxy]propionate
IUPAC navn oxolan-2-ylmethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
SMIL CC(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC4CCCO4
Tekniske data
Kemisk navn eller materiale Quizalofop-p-tefuryl
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C22H21ClN2O5/c1-14(22(26)28-13-18-3-2-10-27-18)29-16-5-7-17(8-6-16)30-21-12-24-20-11-15(23)4-9-19(20)25-21/h4-9,11-12,14,18H,2-3,10,13H2,1H3
Formel vægt 428.1139
Procent renhed >95%