Få mere at vide
Quintrione, TRC
Artikelnummer. 30137896
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137896 250 mg
30136584 500 mg
30008271 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137896 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ135090250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1350901-36-8
Molekylær formel C16H11Cl2NO3
Molekylvægt (g/mol) 336.17
Synonym Quintrione, 1,3-Cyclohexanedione, 2-[(3,7-dichloro-8-quinolinyl)carbonyl]-, 2-[(3,7-Dichloro-8-quinolinyl)carbonyl]-1,3-cyclohexanedione, Dichloroquinolinone, Quintrione, 2-[(3,7-Dichloro-8-quinolinyl)carbonyl]-1,3-cyclohexanedione, Dichloroquinolinone;
IUPAC navn 2-(3,7-dichloroquinoline-8-carbonyl)cyclohexane-1,3-dione
SMIL Clc1cnc2c(C(=O)C3C(=O)CCCC3=O)c(Cl)ccc2c1
Tekniske data
Kemisk navn eller materiale Quintrione
Mængde 250 mg
InChI formel InChI=1S/C16H11Cl2NO3/c17-9-6-8-4-5-10(18)13(15(8)19-7-9)16(22)14-11(20)2-1-3-12(14)21/h4-7,14H,1-3H2
Formel vægt 335.0116