Få mere at vide
Quinoxaline-5,6,7,8-d4, CDN
Artikelnummer. 30317401
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317401 0.5 g
30309019 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317401 Leverandør CDN Leverandørnr. CDND10330.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quinoxaline-5,6,7,8-D4
CAS 64252-08-0
Molekylær formel C8 D4 H2 N2
Mængde 0.5 g
Synonym Quinoxaline-5,6,7,8-d4 (9 CI), 1,4-Benzodiazine-5,6,7,8-d4, 1,4-Diazanaphthalene--5,6,7,8-d4, Quinoxaline-d4
InChI formel InChI=1 S/C8H6N2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-6 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c2nccnc2c1[2 H]
IUPAC navn 5,6,7,8-tetradeuterioquinoxaline
Molekylvægt (g/mol) 134.1713
Formel vægt 134.0782 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity