missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Quinolinium dichromate, 97%

Brand:  Alfa Aesar™ H55300.06

Additional Details : CAS Number : 56549-24-7 Weight : 0.00500kg Transport : UN number : 3087 Chem class : 5.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15491508

  • 279.00 DKK / 5g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

56549-24-7
476.325
PYEIMZCGCWSHSV-UHFFFAOYSA-P
11465689
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
C18H16Cr2N2O7
MFCD00059108
quinolinium dichromate, acmc-209lt3
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium

Specifications

Quinolinium dichromate
164°C to 167°C (decomposition)
56549-24-7
MFCD00059108
quinolinium dichromate, acmc-209lt3
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
476.325
476.33
97%
Orange to Yellow
5g
C18H16Cr2N2O7
UN3087
PYEIMZCGCWSHSV-UHFFFAOYSA-P
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
11465689
Powder
Videos
SDS Documents