Få mere at vide
Quinoline Yellow, TRC
Artikelnummer. 30133836
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133836 10 g
30136682 5 g
17942615 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133836 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ70104010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 8003-22-3
Molekylær formel C18H11NO2
Molekylvægt (g/mol) 269.3
Synonym C.I. Solvent Yellow 33, 1,3-Isobenzofurandione, reaction products with dimethylquinoline and methylquinoline, Arlosol Yellow S, C.I. 47000, Chinoline Yellow D Sol.in Spirits, Chinoline Yellow ZSS, D & C Yellow 11, D and C Yellow No. 11, D&C Yellow No. 11, Japan Yellow 204, Japan Yellow No. 204, Kayaset Yellow 2G, Nitro Fast Yellow SL, Oil Yellow SIS, Oplas Yellow 136, Petrol Yellow C, Polysolve Yellow 33G, Quinoline Yellow A Spirit Soluble, Quinoline Yellow Base, Quinoline Yellow SS, Quinoline Yellow Spirit Soluble, Solvent Yellow 33, Waxoline Yellow T, Yellow 11, C.I. 47000
IUPAC navn 2-(6-methyl-1H-quinolin-2-ylidene)indene-1,3-dione
SMIL Cc1ccc2NC(=C3C(=O)c4ccccc4C3=O)C=Cc2c1
Tekniske data
Kemisk navn eller materiale Solvent Yellow 33
Mængde 10 g
InChI formel InChI=1S/C19H13NO2/c1-11-6-8-15-12(10-11)7-9-16(20-15)17-18(21)13-4-2-3-5-14(13)19(17)22/h2-10,20H,1H3
Formel vægt 287.0946