Få mere at vide
Quinoline-d7, TRC
Artikelnummer. 30427115
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30427115 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427115 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ70109025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quinoline-2,3,4,5,6,7,8-D7
CAS 34071-94-8
Renhedsgrad noter GC
Molekylær formel C9 2H7 N
Mængde 25 mg
Synonym Quinoline D7 (8CI,9CI), Quinoline-2,3,4,5,6,7,8-D7
InChI formel InChI=1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]c1nc2c([2H])c([2H])c([2H])c([2H])c2c([2H])c1[2H]
IUPAC navn 2,3,4,5,6,7,8-heptadeuterioquinoline
Molekylvægt (g/mol) 136.2
Formel vægt 136.1018
Procent renhed >95