Få mere at vide
Quinine Sulfate Dihydrate, TRC
Artikelnummer. 30132668
Skift visning
Mængde:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132668 25 g
30138770 250 g
17932715 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132668 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ69400125G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 207671-44-1
Molekylær formel 2 C20 H24 N2 O2 . 2 H2 O . H2 O4 S
Molekylvægt (g/mol) 782.94
Synonym Quinine sulfate, bis[(R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol] sulfate, Quinine Sulfate Dihydrate, Cinchonan-9-ol, 6'-methoxy-, (8α,9R)-, sulfate, hydrate (2:1:2) (ACI) H411, H410, H361d, H361, H341, H335, H334, H332, H319, H317, H315, H312, 302+H312+H332, H302, Cinchonan-9-ol, 6'-methoxy-, (8α,9R)-, sulfate (2:1) (salt), monohydrate (9CI)
IUPAC navn (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
SMIL O.O.COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)[C@@H]7C[C@@H]8CCN7C[C@@H]8C=C)c6c5.OS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Quinine sulfate dihydrate
Mængde 25 g
InChI formel InChI=1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1
Formel vægt 782.3561