missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Quinine Dihydrochloride, Mikromol™
Description
Quinine Dihydrochloride
Specifications
Specifications
| Kemisk navn eller materiale | Quinine Dihydrochloride |
| CAS | 60-93-5 |
| Analyt- eller komponentnavne | Quinine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H24 N2 O2 . 2 Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Quinine Dihydrochloride, (-)-Quinine dihydrochloride, Acid quinine hydrochloride, Quinine bimuriate, (8alpha,9R)-6'-Methoxycinchonan-9-ol hydrochloride (1:2), (8alpha,9R)-6'-Methoxycinchonan-9-ol dihydrochloride, Quinidine Sulfate Impurity A as Dihydrochloride, Quinidine Impurity A as Dihydrochloride |
| InChI formel | InChI=1S/C20H24N2O2.2ClH/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;2*1H/t13-,14-,19-,20+;;/m0../s1 |
| SMIL | Cl.Cl.COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1 |
| IUPAC navn | (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?