Få mere at vide
Quinine Dihydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Quinine Dihydrochloride |
| CAS | 60-93-5 |
| Analyt- eller komponentnavne | Quinine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H24 N2 O2 . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Quinine Dihydrochloride, (-)-Quinine dihydrochloride, Acid quinine hydrochloride, Quinine bimuriate, (8alpha,9R)-6'-Methoxycinchonan-9-ol hydrochloride (1:2), (8alpha,9R)-6'-Methoxycinchonan-9-ol dihydrochloride, Quinidine Sulfate Impurity A as Dihydrochloride, Quinidine Impurity A as Dihydrochloride |
| InChI formel | InChI=1S/C20H24N2O2.2ClH/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;2*1H/t13-,14-,19-,20+;;/m0../s1 |
| Løsningstype | Neat |
| SMIL | Cl.Cl.COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1 |
| Vis mere |