Få mere at vide
Quinethazone, LoGiCal
Artikelnummer. 30459121
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459121 10 mg
1 options
Artikelnummer. 30459121 Leverandør LoGiCal Leverandørnr. LGCFOR1879.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quinethazone
CAS 73-49-4
Analyt- eller komponentnavne Quinethazone
Forsendelsestilstand Room Temperature
Molekylær formel C10 H12 Cl N3 O3 S
Certificeringer/compliance ISO 9001
Synonym 6-Quinazolinesulfonamide, 7-chloro-2-ethyl-1,2,3,4-tetrahydro-4-oxo-, 2-Ethyl-7-chloro-1,2,3,4-tetrahydro-4-oxoquinazoline-6-sulfonamide, 2-Ethyl-7-chloro-2,3-dihydro-4(1H)-quinazolone-6-sulfonamide, 7-Chloro-2-ethyl-1,2,3,4-tetrahydro-4-oxo-6-quinazolinesulfonamide, 7-Chloro-2-ethyl-1,2,3,4-tetrahydro-4-oxo-6-sulfamoylquinazoline, 7-Chloro-2-ethyl-2,3-dihydro-4-oxo-(1H)-quinazoline-6-sulfonamide, 7-Chloro-2-ethyl-6-sulfamyl-1,2,3,4-tetrahydro-4-quinazolinone, Aquamox, CL 36010, Hydromox, Idrokin, Quimethazin, Quinethazon, Quinethazone, Quinethazonum
InChI formel InChI=1S/C10H12ClN3O3S/c1-2-9-13-7-4-6(11)8(18(12,16)17)3-5(7)10(15)14-9/h3-4,9,13H,2H2,1H3,(H,14,15)(H2,12,16,17)
Løsningstype Neat
SMIL CCC1NC(=O)c2cc(c(Cl)cc2N1)S(=O)(=O)N
IUPAC navn 7-chloro-2-ethyl-4-oxo-2,3-dihydro-1H-quinazoline-6-sulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 289.74
Mængde 10 mg
Formel vægt 289.0288 g/mol