Få mere at vide
Quinaprilat, TRC
Artikelnummer. 30133249
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133249 25 mg
17930705 5 mg
17951195 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133249 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ67001025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82768-85-2
Molekylær formel C23 H26 N2 O5
Molekylvægt (g/mol) 410.46
Synonym 3-Isoquinolinecarboxylic acid, 2-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-, (3S)-, 3-Isoquinolinecarboxylic acid, 2-[2-[(1-carboxy-3-phenylpropyl)amino]-1-oxopropyl]-1,2,3,4-tetrahydro-, [3S-[2[R*(R*)],3R*]]-, CI 928, Quinaprilat, Quinapril Hydrochloride Imp. C (EP)
IUPAC navn (3S)-2-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMIL C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2Cc3ccccc3C[C@H]2C(=O)O
Tekniske data
Kemisk navn eller materiale Quinaprilat
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C23H26N2O5/c1-15(24-19(22(27)28)12-11-16-7-3-2-4-8-16)21(26)25-14-18-10-6-5-9-17(18)13-20(25)23(29)30/h2-10,15,19-20,24H,11-14H2,1H3,(H,27,28)(H,29,30)/t15-,19-,20-/m0/s1
Formel vægt 410.1842
Procent renhed >95%