Få mere at vide
Quinaprilat-d5, TRC
Artikelnummer. 30415194
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30415194 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415194 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ67001210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quinaprilat-d5
CAS 1279034-23-9
Molekylær formel C23 D5 H21 N2 O5
Mængde 10 mg
Synonym Quinaprilat D5
InChI formel InChI=1S/C23H26N2O5/c1-15(24-19(22(27)28)12-11-16-7-3-2-4-8-16)21(26)25-14-18-10-6-5-9-17(18)13-20(25)23(29)30/h2-10,15,19-20,24H,11-14H2,1H3,(H,27,28)(H,29,30)/t15-,19-,20-/m0/s1/i2D,3D,4D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](C)C(=O)N2Cc3ccccc3C[C@H]2C(=O)O)C(=O)O)c([2H])c1[2H]
IUPAC navn (3S)-2-[(2S)-2-[[(1S)-1-carboxy-3-(2,3,4,5,6-pentadeuteriophenyl)propyl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
Molekylvægt (g/mol) 415.494
Formel vægt 415.216