Få mere at vide
Quinapril-d5 Hydrochloride (Major), TRC
Artikelnummer. 30416746
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416746 1 mg
30421767 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416746 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ6700021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quinapril-d5 Hydrochloride
CAS 1356020-03-5
Molekylær formel C25H26D5ClN2O5
Mængde 1 mg
Synonym CI-906-d5','PD-109452-2-d5','Accupril-d5','Accuprin-d5','Accupro-d5, Korec-d5, Korectic-d5
InChI formel InChI=1S/C25H30N2O5.ClH/c1-3-32-25(31)21(14-13-18-9-5-4-6-10-18)26-17(2)23(28)27-16-20-12-8-7-11-19(20)15-22(27)24(29)30;/h4-12,17,21-22,26H,3,13-16H2,1-2H3,(H,29,30);1H/t17?,21-,22+;/m1./s1/i4D,5D,6D,9D,10D;
SMIL O=C(C(N[C@H](CCC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C(OCC)=O)C)N2CC3=C[C]=CC=C3C[C@H]2C(O)=O.Cl
Molekylvægt (g/mol) 480.01