Få mere at vide
Quinalphos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Quinalphos |
| CAS | 13593-03-8 |
| Molekylær formel | C12 H15 N2 O3 P S |
| Mængde | 500 mg |
| Synonym | BAY 77049, Bayer 77049, Bayrusil, Chinalphos, Diethchinalphion, Diethquinalphion, Diethyl 2-quinoxalyl phosphorothionate, Ekalux, Ekalux 25EC, Ekalux G5, Kinalux, O,O-Diethyl O-(2-quinoxalyl) phosphorothionate, O,O-Diethyl O-(quinoxalin-2-yl) thiophosphate, O,O-Diethyl O-2-quinoxalinyl phosphorothioate, O,O-Diethyl-O-(2-quinoxalyl)thiophosphate, O,O-Diethyl-O-quinoxal-2-yl thionophosphate, Oleoekalux, Quinalfos, Quinalphos, Quinaltaf, SAN 6538, SAN 6626, Sandoz 6538, Sandoz 6626 |
| InChI formel | InChI=1S/C12H15N2O3PS/c1-3-15-18(19,16-4-2)17-12-9-13-10-7-5-6-8-11(10)14-12/h5-9H,3-4H2,1-2H3 |
| SMIL | CCOP(=S)(OCC)Oc1cnc2ccccc2n1 |
| IUPAC navn | diethoxy-quinoxalin-2-yloxy-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 298.3 |
| Formel vægt | 298.0541 |