Learn More
Quinaldic Acid Sodium Salt, TRC
Product Code. 30418142
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418142 2.5 g 2.5g
30409752 500 mg 500mg
30409608 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30418142 Supplier Toronto Research Chemicals Supplier No. TRCQ5850082.5G
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16907-79-2
Molecular Formula C10H6NNaO2
Molecular Weight (g/mol) 195.15
IUPAC Name sodium;quinoline-2-carboxylate
SMILES [Na+].[O-]C(=O)c1ccc2ccccc2n1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C10H7NO2.Na/c12-10(13)9-6-5-7-3-1-2-4-8(7)11-9;/h1-6H,(H,12,13);/q;+1/p-1
Formula Weight 195.0296
Chemical Name or Material Quinaldic Acid Sodium Salt