Få mere at vide
Quinacrine Dihydrochloride, TRC
Artikelnummer. 30423912
Skift visning
Mængde:
50 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423912 500 mg
30421968 10 g
30411283 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423912 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ550610500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 69-05-6
Molekylær formel C23 H30 Cl N3 O . 2 Cl H
Molekylvægt (g/mol) 472.88
Synonym Mepacrine Hydrochloride, 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, hydrochloride (1:2), 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, dihydrochloride (9CI), Acridine, 6-chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxy-, dihydrochloride (8CI), 2-Chloro-5-(ω-diethylamino-α-methylbutylamino)-7-methoxyacridine dihydrochloride, 2-Methoxy-6-chloro-9-(4-diethylamino-1-methylbutylamino)acridine dihydrochloride, 3-Chloro-7-methoxy-9-(1-methyl-4-diethylaminobutylamino)acridine dihydrochloride, 3-Chloro-9-(4'-diethylamino-1'-methylbutylamino)-7-methoxyacridine dihydrochloride, 6-Chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxyacridine dihydrochloride, 866RP, Acrichin dihydrochloride, Atabrine hydrochloride, Atebrin, Atebrine, Atebrine hydrochloride, Chemiochin, Chinacrin hydrochloride, Crinodora, Erion, Malaricida, Mecryl, Mepacrine dihydrochloride, Mepacrine hydrochloride, Methoquine, Metochin, Metoquin, Metoquine, NSC 14229, Palacrin, Palusan, Pentilen, Quinacrine dihydrochloride, Quinacrine hydrochloride, SN 390
IUPAC navn 4-N-(6-chloro-2-methoxyacridin-9-yl)-1-N,1-N-diethylpentane-1,4-diamine;dihydrochloride
SMIL Cl.Cl.CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13
Tekniske data
Kemisk navn eller materiale Mepacrine Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H
Formel vægt 471.1611