Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
Brand: Quickfit MF17/3/SC
1282.50 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
- With sidearm for the attachment of rubber hose etc
- Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Specifications
NS 24/29 | |
With Side-Arm for the Attachment of Rubber Hose | |
Tubing Adapter | |
NS 24/29 |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14