Learn More
Quickfit™ PTFE Clamp
For retaining glassware having Quickfit™ conical joints
Brand:  Quickfit GC14
310.65 DKK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 11941528
  • 327.00 DKK / Pack of 5
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Used for retaining glassware having Quickfit™ conical joints
  • Manufactured from PTFE with encapsulated steel spring to provide firm grip
  • Clips are numbered with joint size for easy identification
  • Can be used at temperatures up to 250°C
Specifications
Specifications
Joint Clamp
Encapsulated Steel spring
14/23
PTFE
Conical Joints
250°C