Learn More
Quickfit™ Polyacetal Clamp for Spherical Joints
For retaining glassware having Quickfit™ spherical joints
376.00 DKK - 427.00 DKK
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
12613032
|
Quickfit
KS13 |
Violet |
376.00 DKK
Pack of 5 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||||
12623032
|
Quickfit
KS35 |
Light Orange |
427.00 DKK
Pack of 5 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||||
12989383
|
Quickfit
KS29 |
Dark Red |
415.00 DKK
Pack of 5 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||||
Description
- Manufactured from polyacetal
- Can considerably reduce the need for clamping
- Easy to fit and remove
- Can be used at temperatures up to 80°C
- Clips are color coded for easy identification
Specifications
5/Pk. | |
Joint Clamp | |
Polyacetal |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Quickfit™ Polyacetal Clamp for Spherical Joints