Learn More
Quickfit™ Spare O Ring for PTFE Stirrer Gland
Product Code. 12959413
Change view
Click to view available options
For Use With (Equipment):
PTFE stirrer gland
Unit Size:
Pack of 10
Product Code. For Use With (Equipment) Unit Size
12959413 PTFE stirrer gland Pack of 10
1 options
Product Code. 12959413 Supplier Quickfit Supplier No. ST4R
  • ONLINE EXCLUSIVE PRICE
    226.00 DKK
    186.33 DKK / Pakke med 10
    Spare 39.67 DKK
    17% Slukket
Forventet forsendelse: 11-01-2027
For use with PTFE Stirrer Gland
  • For use with PTFE stirrer glands
  • Fits 6mm stirrer shaft
  • Supplied in packs of 10
Specifications
For Use With (Equipment) PTFE stirrer gland
Product Type O-Ring
Size 6 mm