Learn More
Quickfit™ Glass Funnel with Rotaflo™ Stopcock and Open Top
With open top instead of socket for easy addition of solutions
Brand: Quickfit D3/22
Additional Details : Weight : 0.18000kg
Description
- Complies with ISO 4800
- With stem inside the cone
- Enables easy fitting to the top of Quickfit™ chromatography columns etc. , where a protruding stem would be undesirable
- GP Rotaflo™ stopcock with PTFE key for grease free performance
- With open top instead of socket for easy addition of solutions
- Economical price
Specifications
Dropping funnel | |
100 mL | |
ISO 4800 | |
GP Rotaflo™ stopcock, PTFE key |
39mm | |
Pyrex Glass, Borosilicate Glass | |
19/26 |