Learn More
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Complies with BS 2735
Brand:  Quickfit FR1L/3UM
 View more versions of this product
Product Code. 15547860
  • 1715.00 DKK / Pack of 6
Only null left
Estimated Shipment: 24-10-2025
Description
Complies with BS 2735
  • With a longer neck than standard Quickfit™ flasks
  • Useful for reducing problems associated with frothing and splashing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 mL
Useful for reducing problems associated with frothing and splashing
Medium
Borosilicate Glass
205 mm
24/29
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask > Capacity: 1000mL