Learn More
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Complies with BS 2735
Brand:  Quickfit FR250/3M
 View more versions of this product
Product Code. 12578895
  • 441.00 DKK / Pack of 2
Only null left
Estimated Shipment: 02-09-2025
Description
Complies with BS 2735
  • With a longer neck than standard Quickfit™ flasks
  • Useful for reducing problems associated with frothing and splashing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL
Useful for reducing problems associated with frothing and splashing
Medium
Borosilicate Glass
165 mm
24/29
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask > Capacity: 250mL