Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Designed to support Raschig rings within plain fractionating columns.
Brand: Quickfit EX13/26
This item is not returnable.
View return policy
Description
- Support is passed through socket of column and rests at the constriction above the cone
- Manufactured from chemically resistant borosilicate glass
Specifications
Designed to support Raschig rings within plain fractionating columns | |
Column packing support | |
Pyrex Borosilicate Glass | |
40/38 socket size |
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Column Packing Support > Fits 40/38 socket size
Spot an opportunity for improvement?Share a Content Correction